Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164706.1 | NP_001158178.1 | 2 | 270 | ||
NM_001164707.1 | NP_001158179.1 | 1 | 323 | |||
NM_212494.2 | NP_997659.2 | 2 | 323 | |||
XM_006256120.2 | XP_006256182.1 | 3 | 354 | |||
GenBank mRNA | BP501235.1 | - | 1 | 337 | ||
CB586341.1 | - | 2 | 270 | |||
DV717217.1 | - | 1 | 295 | |||
FQ220708.1 | - | 2 | 557 |