Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212525.1 | NP_997690.1 | 1, 2 | 78 | ||
XM_008759166.1 | XP_008757388.1 | 1, 2 | 150 | |||
GenBank mRNA | AY247021.1 | - | 1, 2 | 78 | ||
FQ211365.1 | - | 1, 2 | 84 |