Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165936.1 | NP_001159408.1 | 2 | 340 | ||
NM_212532.2 | NP_997697.1 | 2 | 387 | |||
XM_008772714.1 | XP_008770936.1 | 3 | 486 | |||
XM_008772715.1 | XP_008770937.1 | 2 | 649 | |||
GenBank mRNA | BC086443.1 | - | 2 | 288 | 7303579 | |
BC097930.1 | - | 2 | 377 | 7388716 | ||
CO385374.1 | - | 2 | 387 | |||
EV770973.1 | - | 2 | 340 |