Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212546.4 | NP_997711.1 | 2 | 1244 | ||
GenBank mRNA | BC108294.1 | - | 1 | 1106 | 7111250 | |
BC108297.1 | - | 2 | 1244 | 7115041 |