Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOPEY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199942.1 | NP_001186871.1 | 21 | 4380 | ||
NM_015018.3 | NP_055833.2 | 21 | 4407 | |||
XM_011535619.2 | XP_011533921.1 | 20 | 4372 | |||
XM_017010559.1 | XP_016866048.1 | 21 | 4402 | |||
XM_017010560.1 | XP_016866049.1 | 20 | 4330 | |||
XM_017010561.1 | XP_016866050.1 | 21 | 4354 | |||
XM_017010562.1 | XP_016866051.1 | 20 | 4303 | |||
XM_017010563.1 | XP_016866052.1 | 21 | 4381 | |||
XM_017010564.1 | XP_016866053.1 | 21 | 4381 | |||
XM_017010565.1 | XP_016866054.1 | 21 | 4375 | |||
XM_017010566.1 | XP_016866055.1 | 21 | 4375 | |||
XM_017010567.1 | XP_016866056.1 | 20 | 4303 | |||
XM_017010568.1 | XP_016866057.1 | 20 | 4372 | |||
XM_017010569.1 | XP_016866058.1 | 19 | 4289 | |||
XM_017010570.1 | XP_016866059.1 | 20 | 4303 | |||
XM_017010571.1 | XP_016866060.1 | 21 | 4402 | |||
XM_017010572.1 | XP_016866061.1 | 21 | 4402 | |||
XM_017010573.1 | XP_016866062.1 | 21 | 4402 | |||
XM_017010574.1 | XP_016866063.1 | 21 | 4402 | |||
GenBank mRNA | AB029040.2 | - | 4 | 1428 | ||
AK027030.1 | - | 1 | 174 | |||
BC144544.1 | - | 21 | 4278 | 9053071 | ||
BC150619.1 | - | 21 | 4278 | 9056989 | ||
BX648667.1 | - | 21 | 4405 |