Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166300.1 | NP_001159772.1 | 3 | 271 | ||
NM_213567.2 | NP_998732.1 | 2 | 189 | |||
GenBank mRNA | FM113828.1 | - | 3 | 270 |