Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213596.2 | NP_998761.2 | 10 | 3053 | ||
XM_011537922.2 | XP_011536224.1 | 10 | 3242 | |||
XM_017018818.1 | XP_016874307.1 | 8 | 4028 | |||
GenBank mRNA | AK093286.1 | - | 3 | 1295 | ||
AK131519.1 | - | 6 | 2660 | |||
BC146825.1 | - | 10 | 3054 | 8860453 |