Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1302996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213610.3 | NP_998775.2 | 6 | 1138 | ||
XM_006255918.2 | XP_006255980.1 | 6 | 1156 | |||
XM_008772712.1 | XP_008770934.1 | 6 | 1027 | |||
XM_008772713.1 | XP_008770935.1 | 6 | 1015 | |||
GenBank mRNA | BC088194.1 | - | 6 | 1138 | 7373688 | |
FQ225840.1 | - | 5 | 921 |