Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213727.2 | NP_998892.1 | 2 | 4002 | ||
GenBank mRNA | AK034979.1 | - | 1 | 3874 | ||
BC070435.1 | - | 2 | 3955 | 6853754 |