Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214459.2 | NP_999624.1 | 3, 4 | 438 | ||
GenBank mRNA | AY500157.1 | - | 3, 4 | 438 |