Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271427.1 | NP_001258356.1 | 2 | 175 | ||
NM_001271428.1 | NP_001258357.1 | 2 | 175 | |||
NM_001271429.1 | NP_001258358.1 | 2 | 175 | |||
NM_015047.2 | NP_055862.1 | 2 | 175 | |||
XM_005245787.1 | XP_005245844.1 | 2 | 170 | |||
XM_005245788.1 | XP_005245845.1 | 2 | 170 | |||
GenBank mRNA | AK075563.1 | - | 2 | 138 | ||
AK291618.1 | - | 2 | 138 | |||
BC034589.1 | - | 2 | 168 | 4831005 | ||
BX648627.1 | - | 2 | 154 | |||
BX648708.1 | - | 2 | 133 | |||
D42044.2 | - | 2 | 122 | |||
DB097686.1 | - | 2 | 175 |