Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163540.1 | NP_001157012.1 | 33 | 13103 | ||
NM_001163542.1 | NP_001157014.1 | 34 | 12620 | |||
NM_001163549.1 | NP_001157021.1 | 33 | 12666 | |||
NM_001164203.1 | NP_001157675.1 | 34 | 12607 | |||
NM_011117.2 | NP_035247.2 | 32 | 12569 | |||
NM_201385.2 | NP_958787.2 | 32 | 12504 | |||
NM_201386.2 | NP_958788.2 | 32 | 12565 | |||
NM_201387.3 | NP_958789.3 | 32 | 12594 | |||
NM_201388.2 | NP_958790.2 | 32 | 12556 | |||
NM_201389.2 | NP_958791.2 | 32 | 13088 | |||
NM_201390.2 | NP_958792.2 | 32 | 12503 | |||
NM_201391.2 | NP_958793.2 | 32 | 12651 | |||
NM_201392.2 | NP_958794.2 | 32 | 12477 | |||
NM_201393.2 | NP_958795.2 | 32 | 12594 | |||
NM_201394.2 | NP_958796.2 | 32 | 12651 | |||
GenBank mRNA | AK137706.1 | - | 1 | 760 | ||
AY480033.1 | - | 32 | 12569 | |||
AY480034.1 | - | 32 | 12504 | |||
AY480035.1 | - | 32 | 12565 | |||
AY480036.1 | - | 32 | 12594 | |||
AY480037.1 | - | 32 | 12556 | |||
AY480038.1 | - | 32 | 13088 | |||
AY480039.1 | - | 32 | 12503 | |||
AY480040.1 | - | 32 | 12651 | |||
AY480041.1 | - | 32 | 12477 | |||
AY480042.1 | - | 32 | 12594 | |||
AY480043.1 | - | 32 | 12651 |