Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011535.3 | NP_035665.2 | 4 | 1565 | ||
NM_198052.2 | NP_932169.1 | 4 | 1701 | |||
GenBank mRNA | AF429310.1 | - | 3 | 359 | ||
AK052453.1 | - | 4 | 1780 | |||
AK145436.1 | - | 4 | 1482 | |||
AK157247.1 | - | 3 | 1266 | |||
AK172118.1 | - | 3 | 1268 | |||
BC096551.1 | - | 4 | 1701 | 30547736 | ||
U15567.1 | - | 2 | 170 | |||
U57328.1 | - | 3 | 386 |