Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166494.1 | NP_001159966.1 | 7 | 1507 | ||
NM_001166495.1 | NP_001159967.1 | 8 | 1296 | |||
NM_027061.2 | NP_081337.2 | 8 | 1387 | |||
NM_199419.3 | NP_955451.2 | 7 | 1168 | |||
GenBank mRNA | AK006058.1 | - | 7 | 1455 | ||
AY251601.1 | - | 8 | 1332 | |||
AY251602.1 | - | 7 | 1113 | |||
BC050986.1 | - | 8 | 1263 | |||
BC089476.1 | - | 7 | 1168 | 6708062 |