Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030098.2 | NP_084374.2 | 2 | 167 | ||
GenBank mRNA | AK020595.1 | - | 2 | 111 | ||
AK038708.1 | - | 3 | 483 | |||
AY545655.1 | - | 2 | 145 | |||
BC094892.1 | - | 2 | 121 | 6432787 | ||
BC132632.1 | - | 2 | 100 | 40130909 | ||
BC132634.1 | - | 2 | 99 | 40130911 | ||
BY228312.1 | - | 2 | 170 |