Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145514.5 | NP_663489.4 | 11 | 2061 | ||
GenBank mRNA | AK075743.1 | - | 11 | 1453 | ||
AK082326.1 | - | 12 | 1356 | |||
AK213047.1 | - | 2 | 53 | |||
AK218758.1 | - | 2 | 53 | |||
BC020044.1 | - | 7 | 806 | |||
BC058601.1 | - | 9 | 1057 | |||
BC094233.1 | - | 8 | 945 | |||
BC139336.1 | - | 11 | 1223 | 8862358 | ||
BC139337.1 | - | 11 | 1223 | 8862359 |