Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145514.5 | NP_663489.4 | 14 | 2494 | ||
GenBank mRNA | AK075743.1 | - | 14 | 1886 | ||
AK082326.1 | - | 15 | 1789 | |||
AK139350.1 | - | 1 | 45 | |||
AK167017.1 | - | 3 | 303 | |||
AK167116.1 | - | 3 | 303 | |||
BC020044.1 | - | 10 | 1239 | |||
BC058601.1 | - | 12 | 1490 | |||
BC094233.1 | - | 11 | 1378 | |||
BC139336.1 | - | 14 | 1656 | 8862358 | ||
BC139337.1 | - | 14 | 1656 | 8862359 |