Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153599.3 | NP_705827.2 | 13 | 1520 | ||
GenBank mRNA | AK131948.1 | - | 13 | 1445 | ||
BC025046.1 | - | 6 | 977 | |||
BC085293.1 | - | 5 | 632 | |||
BC132551.1 | - | 6 | 835 | |||
BC132553.1 | - | 6 | 835 |