Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304495.1 | NP_001291424.1 | 1 | 506 | ||
NM_171982.4 | NP_741983.2 | 1 | 506 | |||
XM_005273452.3 | XP_005273509.1 | 1 | 501 | |||
XM_011544453.1 | XP_011542755.1 | 1 | 422 | |||
GenBank mRNA | AB029021.1 | - | 1 | 456 | ||
AF492463.1 | - | 1 | 504 | |||
AK308236.1 | - | 1 | 426 | |||
BC018337.1 | - | 1 | 504 | |||
BC069226.1 | - | 1 | 448 | 6428667 |