Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175247.3 | NP_780456.2 | 7 | 999 | ||
GenBank mRNA | AK028202.1 | - | 7 | 972 | ||
AK048530.1 | - | 7 | 991 | |||
AK173170.1 | - | 7 | 964 | |||
AY528713.1 | - | 7 | 905 | |||
BC062116.1 | - | 7 | 967 | 6817400 | ||
M36516.1 | - | 122 |