Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183019.2 | NP_898840.2 | 10 | 1739 | ||
GenBank mRNA | AK034013.1 | - | 10 | 1703 | ||
AK122448.2 | - | 6 | 794 | |||
AK134629.1 | - | 1702 | ||||
AK138598.1 | - | 10 | 1699 | |||
AK163424.1 | - | 10 | 1721 | |||
AK169957.1 | - | 10 | 1759 | |||
BC055804.1 | - | 10 | 1700 | |||
BC094608.1 | - | 5259 | ||||
BC139371.1 | - | 10 | 1722 | 8862393 | ||
BC139397.1 | - | 10 | 1722 | 8862419 |