Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184045.2 | NP_908934.1 | 12 | 1282 | ||
XM_006229558.2 | XP_006229620.1 | 12 | 1417 | |||
XM_006229559.2 | XP_006229621.1 | 12 | 1413 | |||
XM_006229560.2 | XP_006229622.1 | 12 | 1408 | |||
XM_006229561.2 | XP_006229623.1 | 12 | 1405 | |||
XM_008773617.1 | XP_008771839.1 | 10 | 1219 | |||
XM_008773618.1 | XP_008771840.1 | 10 | 1209 | |||
GenBank mRNA | AY346151.1 | - | 12 | 1286 |