Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015074.3 | NP_055889.2 | 2 | 326 | ||
NM_183416.3 | NP_904325.2 | 2 | 326 | |||
GenBank mRNA | AB011163.2 | - | 2 | 275 | ||
AB017133.1 | - | 1 | 158 | |||
AB040881.2 | - | 2 | 385 | |||
AB088210.1 | - | 1 | 74 | |||
AB088211.1 | - | 1 | 74 | |||
AB088212.1 | - | 1 | 74 | |||
AB088213.1 | - | 1 | 74 | |||
AF257176.1 | - | 2 | 227 | |||
AK308121.1 | - | 2 | 326 | |||
AY043362.1 | - | 1 | 78 | |||
AY139835.1 | - | 2 | 227 | |||
BC115395.2 | - | 1 | 162 | 40067993 | ||
BF940695.1 | - | 2 | 259 |