Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243166.1 | NP_001230095.1 | 1 | 227 |
Gene Symbol: Gm13271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085528.1 | NP_001078997.1 | 1 | 67 |
Gene Symbol: Gm13272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161608.1 | NP_001155080.1 | 1 | 67 |
Gene Symbol: Gm13275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085533.2 | NP_001079002.1 | 1 | 812 |
Gene Symbol: Gm13276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085532.3 | NP_001079001.1 | 1 | 722 |
Gene Symbol: Gm13277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098840.3 | NP_001092310.1 | 1 | 722 |
Gene Symbol: Gm13278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098841.3 | NP_001092311.1 | 1 | 722 |
Gene Symbol: Gm13283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085531.2 | NP_001079000.1 | 1 | 67 |
Gene Symbol: Gm13285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161609.1 | NP_001155081.1 | 1 | 67 |
Gene Symbol: Gm13286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243150.1 | NP_001230079.1 | 1 | 67 |
Gene Symbol: Gm13287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161610.1 | NP_001155082.1 | 1 | 67 |
Gene Symbol: Gm13288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243167.1 | NP_001230096.1 | 1 | 722 | ||
GenBank mRNA | AK031317.1 | - | 1 | 723 |
Gene Symbol: Gm13289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243165.1 | NP_001230094.1 | 227 |
Gene Symbol: Gm13290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243155.1 | NP_001230084.1 | 1 | 227 | ||
GenBank mRNA | AB024521.1 | - | 1 | 227 |
Gene Symbol: Ifnz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197889.2 | NP_922871.1 | 1 | 722 | ||
GenBank mRNA | AK046550.1 | - | 1 | 723 |