Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159419.1 | NP_001152891.1 | 5 | 906 | ||
NM_198034.3 | NP_932151.2 | 5 | 906 | |||
GenBank mRNA | AK046836.1 | - | 5 | 1266 | ||
AK171920.1 | - | 5 | 1059 | |||
BC079584.1 | - | 5 | 906 | 5704672 |