Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198731.2 | NP_942026.1 | 8 | 2787 | ||
XM_006252578.2 | XP_006252640.1 | 8 | 2852 | |||
XM_006252579.2 | XP_006252641.1 | 8 | 2806 | |||
XM_006252580.2 | XP_006252642.1 | 8 | 2612 | |||
XM_008770984.1 | XP_008769206.1 | 8 | 3155 | |||
XM_008770985.1 | XP_008769207.1 | 9 | 3244 | |||
GenBank mRNA | BC085787.1 | - | 8 | 2787 | 7108667 |