Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198747.1 | NP_942042.1 | 3 | 1063 | ||
XM_006238245.2 | XP_006238307.1 | 3 | 1113 | |||
GenBank mRNA | AY232999.1 | - | 3 | 1063 |