Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198752.3 | NP_942047.2 | 18 | 2837 | ||
GenBank mRNA | AJ250329.1 | - | 18 | 2838 | ||
AJ250330.1 | - | 17 | 2879 | |||
FQ211833.1 | - | 16 | 2801 |