Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198756.2 | NP_942051.1 | 6 | 491 | ||
GenBank mRNA | BC058507.1 | - | 6 | 495 | 6889538 | |
FQ215521.1 | - | 7 | 670 |