Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161483.1 | NP_001154955.1 | 11 | 2426 | ||
NM_199024.2 | NP_950189.2 | 10 | 3913 | |||
GenBank mRNA | AK029800.1 | - | 6 | 1738 | ||
AK031008.1 | - | 10 | 3913 | |||
AK031753.1 | - | 4 | 1419 | |||
AK081727.1 | - | 1 | 563 | |||
BC056377.1 | - | 11 | 2419 | 6850243 |