Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199084.1 | NP_954515.1 | 13, 14 | 1610 | ||
XM_008770845.1 | XP_008769067.1 | 13, 14 | 1749 | |||
XM_008770846.1 | XP_008769068.1 | 13, 14 | 1831 | |||
XM_008770847.1 | XP_008769069.1 | 13, 14 | 1794 | |||
XM_008770848.1 | XP_008769070.1 | 12, 13 | 1523 | |||
XM_008770849.1 | XP_008769071.1 | 12, 13 | 1666 | |||
XM_008770850.1 | XP_008769072.1 | 12, 13 | 1748 | |||
XM_008770851.1 | XP_008769073.1 | 14, 15 | 1720 | |||
XM_008770852.1 | XP_008769074.1 | 14, 15 | 2717 | |||
XM_008770853.1 | XP_008769075.1 | 14, 15 | 1934 | |||
XM_008770854.1 | XP_008769076.1 | 14, 15 | 1702 | |||
XM_008770855.1 | XP_008769077.1 | 15, 16 | 2098 | |||
XM_008770856.1 | XP_008769078.1 | 12, 13 | 1684 | |||
XM_008770857.1 | XP_008769079.1 | 11, 12 | 1635 | |||
XM_008770858.1 | XP_008769080.1 | 11, 12 | 1590 | |||
XM_008770859.1 | XP_008769081.1 | 10, 11 | 1428 | |||
XM_008770860.1 | XP_008769082.1 | 10, 11 | 1361 | |||
XM_008770861.1 | XP_008769083.1 | 11, 12 | 1503 | |||
GenBank mRNA | BC061766.1 | - | 13, 14 | 1610 | 5621190 | |
FQ211316.1 | - | 11, 12 | 1468 | |||
FQ211373.1 | - | 11, 12 | 1468 |