Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199085.2 | NP_954516.1 | 6, 7 | 849 | ||
XM_006253869.1 | XP_006253931.1 | 6, 7 | 847 | |||
XM_006253870.2 | XP_006253932.1 | 6, 7 | 1142 | |||
XM_006253871.2 | XP_006253933.1 | 6, 7 | 962 | |||
XM_008771596.1 | XP_008769818.1 | 6, 7 | 1060 | |||
GenBank mRNA | BC060594.1 | - | 6, 7 | 853 | 6887824 |