Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199088.1 | NP_954519.1 | 8 | 1314 | ||
XM_006229014.2 | XP_006229076.1 | 8 | 1376 | |||
XM_006229015.2 | XP_006229077.1 | 8 | 1349 | |||
XM_006229016.2 | XP_006229078.1 | 8 | 1237 | |||
XM_006229017.2 | XP_006229079.1 | 8 | 1348 | |||
XM_006229018.2 | XP_006229080.1 | 8 | 1375 | |||
XM_006229019.2 | XP_006229081.1 | 8 | 1375 | |||
XM_006229020.2 | XP_006229082.1 | 8 | 1348 | |||
GenBank mRNA | AY346104.1 | - | 8 | 1314 |