Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Theg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011583.3 | NP_035713.1 | 7 | 927 | ||
NM_199226.1 | NP_954696.1 | 6 | 855 | |||
GenBank mRNA | AB033128.1 | - | 7 | 909 | ||
AJ011834.1 | - | 7 | 1115 | |||
AK017058.1 | - | 6 | 845 | |||
BC050970.1 | - | 6 | 856 | |||
BC061243.1 | - | 6 | 801 | 6775002 |