Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015143.2 | NP_055958.2 | 5 | 537 | ||
XM_011531777.2 | XP_011530079.1 | 5 | 534 | |||
XM_011531778.1 | XP_011530080.1 | 5 | 330 | |||
XM_011531779.1 | XP_011530081.1 | 5 | 537 | |||
XM_017007914.1 | XP_016863403.1 | 5 | 537 | |||
XM_017007915.1 | XP_016863404.1 | 6 | 551 | |||
XM_017007916.1 | XP_016863405.1 | 5 | 550 | |||
XM_017007917.1 | XP_016863406.1 | 5 | 326 | |||
GenBank mRNA | AK304239.1 | - | 5 | 453 | ||
BC030054.1 | - | 1 | 95 | 3958447 | ||
CR936632.1 | - | 7 | 4115 | |||
D42084.1 | - | 5 | 445 |