Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143854.1 | NP_001137326.1 | 5 | 575 | ||
NM_014954.3 | NP_055769.2 | 4 | 563 | |||
XM_011538048.1 | XP_011536350.1 | 5 | 379 | |||
GenBank mRNA | AB023202.1 | - | 5 | 496 | ||
AK295696.1 | - | 5 | 529 | |||
AK316112.1 | - | 5 | 410 | |||
BC002605.2 | - | 3 | 301 | |||
BC009843.2 | - | 3 | 224 | |||
BC017259.2 | - | 4 | 370 | 3510158 | ||
DA332963.1 | - | 5 | 575 |