Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025014.1 | NP_079290.1 | 5 | 1107 | ||
NM_173728.3 | NP_776089.2 | 5 | 1171 | |||
XM_011523734.1 | XP_011522036.1 | 5 | 1171 | |||
XM_011523735.1 | XP_011522037.1 | 5 | 1107 | |||
XM_011523736.1 | XP_011522038.1 | 5 | 1171 | |||
XM_011523737.1 | XP_011522039.1 | 2 | 163 | |||
GenBank mRNA | AB020722.1 | - | 5 | 1087 | ||
AK291626.1 | - | 5 | 1131 | |||
AK300327.1 | - | 5 | 592 | |||
BC036749.1 | - | 5 | 1126 | 5204409 | ||
KU178557.1 | - | 4 | 1016 | |||
KU178558.1 | - | 4 | 1016 |