Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBNO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100122.1 | NP_001093592.1 | 11 | 1327 | ||
NM_014963.2 | NP_055778.2 | 14 | 1659 | |||
XM_011527804.2 | XP_011526106.1 | 14 | 1632 | |||
GenBank mRNA | AB023180.1 | - | 14 | 1636 | ||
AK074102.1 | - | 10 | 1119 | |||
AK125139.1 | - | 11 | 1274 | |||
AK292407.1 | - | 14 | 1659 | |||
AK296849.1 | - | 11 | 1274 | |||
AK300981.1 | - | 15 | 1599 | |||
AK309167.1 | - | 11 | 1274 | |||
BQ950129.1 | - | 4 | 418 |