Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H13 SIRNA Assays ›
Gene Symbol:
ZC3H13
Gene Name:
zinc finger CCCH-type containing 13
Gene Aliases:
KIAA0853
Chromosome Location:
Chr.13: 45954469 - 46052785 on Build GRCh38
UniGene ID:
Hs.136102
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22493
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015070.4
NP_055885.3
10
1395
XM_005266301.3
XP_005266358.1
10
1689
XM_005266302.3
XP_005266359.1
10
1389
XM_005266303.3
XP_005266360.1
11
1714
XM_005266304.3
XP_005266361.1
10
1671
XM_005266305.3
XP_005266362.1
11
1707
XM_005266306.3
XP_005266363.1
10
1385
XM_005266307.3
XP_005266364.1
10
1689
XM_005266308.3
XP_005266365.1
10
1689
XM_005266309.3
XP_005266366.1
10
1389
XM_005266310.3
XP_005266367.1
10
1671
XM_005266311.3
XP_005266368.1
10
1689
XM_005266312.1
XP_005266369.1
10
1689
XM_017020475.1
XP_016875964.1
11
1477
XM_017020476.1
XP_016875965.1
11
1474
XM_017020477.1
XP_016875966.1
11
1714
XM_017020478.1
XP_016875967.1
11
1707
XM_017020479.1
XP_016875968.1
11
1477
XM_017020480.1
XP_016875969.1
10
1385
XM_017020481.1
XP_016875970.1
11
1714
XM_017020482.1
XP_016875971.1
10
1389
GenBank mRNA
AK056422.1
-
10
1672
AK092988.1
-
7
1034
AL136745.1
-
3
431
AL831833.1
-
5
799
AY283618.1
-
10
1393
BC032425.1
-
3
563
BC050658.2
-
7
1064
BC062685.1
-
3
461
BC107864.1
-
3
475
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0