Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015077.3 | NP_055892.2 | 2, 3 | 1457 | ||
GenBank mRNA | AB011096.1 | - | 2, 3 | 709 | ||
AJ290445.1 | - | 2, 3 | 1012 | |||
AK302038.1 | - | 3, 4 | 786 | |||
AK314609.1 | - | 2, 3 | 1457 | |||
AY444166.1 | - | 2, 3 | 1086 | |||
BC040429.1 | - | 2, 3 | 1045 | |||
BC122860.1 | - | 3, 4 | 765 |