Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303420.1 | NP_001290349.1 | 2 | 777 | ||
NM_001303421.1 | NP_001290350.1 | 2 | 291 | |||
NM_015101.3 | NP_055916.1 | 2 | 777 | |||
XM_011509332.2 | XP_011507634.1 | 2 | 484 | |||
XM_011509334.2 | XP_011507636.1 | 2 | 291 | |||
XM_011509335.2 | XP_011507637.1 | 2 | 342 | |||
XM_011509336.2 | XP_011507638.1 | 2 | 119 | |||
XM_017000750.1 | XP_016856239.1 | 2 | 342 | |||
GenBank mRNA | AB011156.1 | - | 2 | 702 | ||
AF288389.1 | - | 2 | 736 | |||
AK095192.1 | - | 2 | 265 | |||
AK293973.1 | - | 2 | 484 | |||
BC035672.1 | - | 2 | 759 | 5740842 | ||
DA216863.1 | - | 2 | 291 | |||
DR761966.1 | - | 2 | 733 |