Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015113.3 | NP_055928.3 | 33 | 5319 | ||
XM_005256561.2 | XP_005256618.1 | 33 | 5383 | |||
XM_017024382.1 | XP_016879871.1 | 33 | 5383 | |||
XM_017024383.1 | XP_016879872.1 | 33 | 5380 | |||
XM_017024384.1 | XP_016879873.1 | 8 | 1194 | |||
GenBank mRNA | AB007859.4 | - | 33 | 5319 | ||
AK001683.1 | - | 4 | 418 | |||
AK122719.1 | - | 8 | 1413 | |||
BC151836.1 | - | 33 | 5319 | 8860131 |