Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015138.4 | NP_055953.3 | 3, 4 | 477 | ||
GenBank mRNA | AK292302.1 | - | 3, 4 | 449 | ||
BC015052.2 | - | 3, 4 | 427 | 3909737 | ||
BF979715.1 | - | 3, 4 | 477 | |||
D87440.2 | - | 3, 4 | 443 |