Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 4 | 395 | ||
NM_001322494.1 | NP_001309423.1 | 4 | 462 | |||
NM_001322495.1 | NP_001309424.1 | 4 | 1224 | |||
GenBank mRNA | AK293144.1 | - | 4 | 286 | ||
AK298292.1 | - | 3 | 270 | |||
AK311421.1 | - | 4 | 366 | |||
AK311567.1 | - | 3 | 177 | |||
AK312492.1 | - | 4 | 375 | |||
BC007927.1 | - | 4 | 382 | 3946317 | ||
DC328543.1 | - | 4 | 462 | |||
EU668322.1 | - | 4 | 382 | |||
EU794618.1 | - | 4 | 382 | |||
KU178105.1 | - | 3 | 296 | |||
KU178106.1 | - | 3 | 296 | |||
U34683.1 | - | 4 | 336 |