Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000214.2 | NP_000205.1 | 2, 3 | 913 | ||
GenBank mRNA | AF003837.1 | - | 2, 3 | 856 | ||
AF028593.1 | - | 2, 3 | 660 | |||
AK312722.1 | - | 2, 3 | 482 | |||
BC098393.1 | - | 2, 3 | 816 | 30528888 | ||
BC126205.1 | - | 3, 4 | 421 | 8991921 | ||
BC126207.1 | - | 2, 3 | 649 | 8991923 | ||
U61276.1 | - | 2, 3 | 767 | |||
U73936.1 | - | 2, 3 | 810 | |||
U77720.1 | - | 2, 3 | 359 |