Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204514.1 | NP_001191443.1 | 15, 16 | 2099 | ||
NM_001204516.1 | NP_001191445.1 | 13, 14 | 1803 | |||
NM_001204517.1 | NP_001191446.1 | 14, 15 | 3044 | |||
NM_014970.3 | NP_055785.2 | 14, 15 | 1935 | |||
XM_005244970.1 | XP_005245027.1 | 14, 15 | 1938 | |||
XM_011509307.2 | XP_011507609.2 | 14, 15 | 1938 | |||
GenBank mRNA | AK026471.1 | - | 14, 15 | 1714 | ||
AK122897.1 | - | 14, 15 | 3046 | |||
AK301912.1 | - | 7, 8 | 902 | |||
AK303052.1 | - | 15, 16 | 2056 | |||
AK315586.1 | - | 14, 15 | 1849 | |||
BC028679.2 | - | 14, 15 | 1892 | 4829354 | ||
BC048012.1 | - | 13, 14 | 1847 | |||
U59919.1 | - | 14, 15 | 1852 |