Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145860.1 | NP_001139332.1 | 7, 8 | 1119 | ||
NM_001145861.1 | NP_001139333.1 | 7, 8 | 1059 | |||
NM_015029.2 | NP_055844.2 | 7, 8 | 1047 | |||
XM_011516801.2 | XP_011515103.1 | 7, 8 | 1119 | |||
GenBank mRNA | AK000837.1 | - | 1, 2 | 113 | ||
AK291434.1 | - | 7, 8 | 1116 | |||
BC011529.1 | - | 7, 8 | 1047 | 3853321 | ||
D31765.1 | - | 4, 5 | 658 |