Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015050.2 | NP_055865.1 | 24 | 3747 | ||
GenBank mRNA | AK025809.1 | - | 5 | 1453 | ||
AK124393.1 | - | 1 | 4305 | |||
BC010731.2 | - | 9 | 1874 | |||
BC031890.1 | - | 24 | 3640 | 4944457 | ||
D43949.2 | - | 24 | 3637 |