Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135608.1 | NP_001129080.1 | 23 | 4840 | ||
NM_015071.4 | NP_055886.1 | 23 | 5005 | |||
XM_005268398.4 | XP_005268455.1 | 23 | 5242 | |||
XM_005268399.4 | XP_005268456.1 | 23 | 5074 | |||
XM_005268400.4 | XP_005268457.1 | 22 | 4964 | |||
XM_005268402.4 | XP_005268459.1 | 22 | 5083 | |||
XM_017009247.1 | XP_016864736.1 | 24 | 5319 | |||
XM_017009248.1 | XP_016864737.1 | 24 | 5106 | |||
XM_017009249.1 | XP_016864738.1 | 24 | 5150 | |||
GenBank mRNA | AB014521.1 | - | 22 | 4786 | ||
AK124521.1 | - | 3 | 3238 |