Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 26 | 5123 | ||
XM_011514422.1 | XP_011512724.1 | 25 | 5279 | |||
XM_011514423.2 | XP_011512725.1 | 26 | 5381 | |||
XM_011514424.1 | XP_011512726.1 | 25 | 5198 | |||
XM_011514425.1 | XP_011512727.1 | 23 | 4949 | |||
XM_011514426.1 | XP_011512728.1 | 25 | 5279 | |||
XM_011514427.1 | XP_011512729.1 | 21 | 3898 | |||
XM_011514428.1 | XP_011512730.1 | 25 | 5279 | |||
XM_011514429.1 | XP_011512731.1 | 18 | 3103 | |||
XM_017010589.1 | XP_016866078.1 | 25 | 5990 | |||
XM_017010590.1 | XP_016866079.1 | 19 | 3452 | |||
GenBank mRNA | AB014608.3 | - | 18 | 2757 | ||
AJ318215.1 | - | 25 | 5048 | |||
AY145132.1 | - | 25 | 5048 | |||
BC002879.2 | - | 3 | 389 | |||
CR749511.1 | - | 25 | 5502 | |||
CR749841.1 | - | 26 | 5122 |